Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpdl3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133888.2 | NP_598649.1 | 6 | 950 | ||
GenBank mRNA | AK136602.1 | - | 5 | 612 | ||
AK154599.1 | - | 6 | 976 | |||
AK166253.1 | - | 6 | 926 | |||
BC009087.1 | - | 6 | 950 | 2810972 |