Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc15a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133895.1 | NP_598656.1 | 3 | 912 | ||
GenBank mRNA | AK183378.1 | - | 1 | 55 | ||
AK214917.1 | - | 3 | 402 | |||
AY050630.1 | - | 3 | 924 | |||
BC016233.1 | - | 3 | 912 | 4502184 |
Gene Symbol: Psmd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK180397.1 | - | 102 |