Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsun5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145414.2 | NP_663389.2 | 4 | 476 | ||
GenBank mRNA | AF412029.1 | - | 4 | 469 | ||
AK079400.1 | - | 4 | 477 | |||
AK149521.1 | - | 4 | 458 | |||
AK156651.1 | - | 4 | 471 |