Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pds5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175310.6 | NP_780519.3 | 20 | 2342 | ||
GenBank mRNA | AK041682.1 | - | 6 | 725 | ||
AK045159.1 | - | 3 | 325 | |||
AK086753.1 | - | 2 | 94 | |||
AK122414.1 | - | 13 | 1432 | |||
AY102267.1 | - | 20 | 2342 | |||
BC096539.1 | - | 19 | 2205 | 6405163 |