Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133905.2 | NP_598666.1 | 5 | 705 | ||
GenBank mRNA | AK033141.1 | - | 2 | 42 | ||
AK146086.1 | - | 5 | 689 | |||
AM419011.1 | - | 4 | 478 | |||
BC016629.1 | - | 5 | 705 | 3595512 |