Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133907.3 | NP_598668.1 | 23 | 3438 | ||
GenBank mRNA | AK029973.1 | - | 23 | 3248 | ||
AK046056.1 | - | 23 | 3439 | |||
AK049125.1 | - | 6 | 829 | |||
AK122187.1 | - | 23 | 3373 | |||
BC021525.1 | - | 12 | 1683 | |||
BC059245.1 | - | 23 | 3253 | |||
BC120731.1 | - | 23 | 3185 | 40129654 | ||
BC137626.1 | - | 23 | 3185 | 8860645 |