Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emilin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133918.2 | NP_598679.1 | 4 | 1898 | ||
GenBank mRNA | AK020015.1 | - | 2 | 1598 | ||
AK029337.1 | - | 4 | 1869 | |||
AK052368.1 | - | 4 | 1979 | |||
AK088995.1 | - | 4 | 1962 | |||
AK138020.1 | - | 2 | 1574 | |||
AK140388.1 | - | 2 | 1314 | |||
AK155486.1 | - | 4 | 2200 | |||
BC005481.1 | - | 4 | 1898 | 3485057 |