Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itfg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133927.1 | NP_598688.1 | 10 | 1083 | ||
GenBank mRNA | AK134630.1 | - | 10 | 1104 | ||
AK147728.1 | - | 10 | 1085 | |||
AK159637.1 | - | 10 | 1092 | |||
BC014833.1 | - | 10 | 1083 | 4507661 | ||
BC025492.1 | - | 10 | 1103 | 5309950 |