Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pot1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133931.4 | NP_598692.1 | 6, 7 | 637 | ||
GenBank mRNA | AK036052.1 | - | 6, 7 | 638 | ||
AK040864.1 | - | 6, 7 | 630 | |||
AK085041.1 | - | 6, 7 | 628 | |||
AK135346.1 | - | 6, 7 | 607 | |||
AK137376.1 | - | 6, 7 | 607 | |||
BC016121.1 | - | 6, 7 | 599 | 4501733 |