Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207210.2 | NP_997093.2 | 10, 11 | 1182 | ||
GenBank mRNA | AK077117.1 | - | 10, 11 | 1183 | ||
BC052324.1 | - | 9, 10 | 1236 | 6315342 |