Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252458.1 | NP_001239387.1 | 17 | 2449 | ||
NM_178900.4 | NP_849231.1 | 16 | 2734 | |||
GenBank mRNA | AK037030.1 | - | 16 | 2735 | ||
BC095949.1 | - | 17 | 2399 | 4950957 | ||
BC096444.1 | - | 17 | 2417 | 4216827 |