Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eftud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159672.1 | NP_001153144.1 | 18 | 2104 | ||
NM_175317.3 | NP_780526.2 | 18 | 2133 | |||
GenBank mRNA | AK028676.1 | - | 18 | 2106 | ||
AK031647.1 | - | 18 | 2090 | |||
BC031852.1 | - | 12 | 1379 | |||
BC045616.1 | - | 18 | 2087 | 4983234 |