Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027256.2 | NP_081532.1 | 21, 22 | 2727 | ||
GenBank mRNA | AK011676.1 | - | 7, 8 | 724 | ||
AK186046.1 | - | 1, 2 | 149 | |||
BC013710.1 | - | 21, 22 | 2673 | 3156446 | ||
BC013813.1 | - | 9, 10 | 981 | |||
BC021838.1 | - | 14, 15 | 1730 | |||
BC031145.1 | - | 15, 16 | 1806 |