Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK083986.1 | - | 1306 | |||
BC046509.1 | - | 893 | ||||
BC094602.1 | - | 992 |