Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133967.3 | NP_598728.1 | 7 | 1133 | ||
GenBank mRNA | AK041905.1 | - | 7 | 1002 | ||
AK089681.1 | - | 979 | ||||
AK090333.1 | - | 7 | 979 | |||
AK157968.1 | - | 7 | 986 | |||
AK170639.1 | - | 7 | 986 | |||
AK170923.1 | - | 7 | 956 | |||
AK172462.1 | - | 7 | 967 | |||
AK213858.1 | - | 4 | 261 | |||
AY166673.1 | - | 5 | 651 | |||
BC013467.1 | - | 6 | 684 | 4218025 | ||
BC029666.1 | - | 2 | 87 | |||
BC075666.1 | - | 7 | 1133 | 30605163 |