Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133967.3 | NP_598728.1 | 8 | 1292 | ||
GenBank mRNA | AK041905.1 | - | 8 | 1161 | ||
AK089681.1 | - | 1138 | ||||
AK090333.1 | - | 8 | 1138 | |||
AK157968.1 | - | 8 | 1145 | |||
AK170639.1 | - | 8 | 1145 | |||
AK170923.1 | - | 8 | 1115 | |||
AK172462.1 | - | 8 | 1126 | |||
AY166673.1 | - | 6 | 810 | |||
BC013467.1 | - | 7 | 843 | 4218025 | ||
BC029666.1 | - | 3 | 246 | |||
BC075666.1 | - | 8 | 1292 | 30605163 |