Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018743.4 | NP_061213.2 | 2 | 1077 | ||
GenBank mRNA | AF406611.1 | - | 1 | 588 | ||
AK045235.1 | - | 2 | 971 | |||
AK049675.1 | - | 2 | 971 | |||
AK077561.1 | - | 2 | 961 | |||
AK083589.1 | - | 2 | 971 | |||
AK161270.1 | - | 2 | 940 | |||
AK167876.1 | - | 2 | 986 | |||
AK169257.1 | - | 2 | 971 | |||
AK178773.1 | - | 1 | 66 | |||
AK182757.1 | - | 1 | 126 | |||
AK188390.1 | - | 1 | 185 | |||
AY489184.1 | - | 1 | 146 | |||
BC031767.1 | - | 1 | 431 | 4457846 |