Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175324.3 | NP_780533.2 | 15 | 1787 | ||
GenBank mRNA | AK077531.1 | - | 15 | 1788 | ||
AK128998.1 | - | 15 | 1766 | |||
AK139918.1 | - | 15 | 1731 | |||
AK145561.1 | - | 15 | 1776 | |||
AK145834.1 | - | 15 | 1767 | |||
BC006042.1 | - | 5 | 463 | |||
BC032994.1 | - | 8 | 716 | |||
BC045199.1 | - | 15 | 1751 | 5133197 | ||
BC110556.2 | - | 6 | 531 |