Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175324.3 | NP_780533.2 | 12 | 1557 | ||
GenBank mRNA | AK077531.1 | - | 12 | 1558 | ||
AK128998.1 | - | 12 | 1536 | |||
AK139918.1 | - | 12 | 1501 | |||
AK145561.1 | - | 12 | 1546 | |||
AK145834.1 | - | 12 | 1537 | |||
BC006042.1 | - | 2 | 233 | |||
BC032994.1 | - | 5 | 486 | |||
BC045199.1 | - | 12 | 1521 | 5133197 |