Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phldb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153537.4 | NP_705765.3 | 6 | 1729 | ||
GenBank mRNA | AK122336.1 | - | 3 | 1447 | ||
AK210349.1 | - | 1 | 207 | |||
BC058712.1 | - | 6 | 1725 | 6842596 |