Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175325.3 | NP_780534.1 | 5 | 370 | ||
GenBank mRNA | AK039560.1 | - | 5 | 319 | ||
AK089970.1 | - | 5 | 325 | |||
AK134734.1 | - | 5 | 323 | |||
BC055797.1 | - | 5 | 290 | |||
BC089507.1 | - | 5 | 304 | |||
BC092531.1 | - | 5 | 314 | |||
BC145771.1 | - | 5 | 332 | 40131074 | ||
BC145773.1 | - | 5 | 332 | 40131076 |