Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D330045A20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175326.5 | NP_780535.1 | 14 | 2641 | ||
GenBank mRNA | AK052376.1 | - | 14 | 2565 | ||
AK155588.1 | - | 14 | 2638 | |||
AK171459.1 | - | 14 | 2637 | |||
BC112416.1 | - | 13 | 2449 | |||
BC113128.1 | - | 14 | 2541 | 40061080 |