Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fig4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133999.1 | NP_598760.1 | 22 | 2717 | ||
GenBank mRNA | AK041196.1 | - | 22 | 2719 | ||
AK172130.1 | - | 22 | 2711 | |||
AK172926.1 | - | 15 | 1765 | |||
BC015295.1 | - | 22 | 2717 | 4221630 | ||
BC031887.1 | - | 22 | 2716 | 5363304 |