Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf3ip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134000.3 | NP_598761.2 | 5 | 1640 | ||
GenBank mRNA | AB238206.1 | - | 4 | 1219 | ||
AB238207.1 | - | 4 | 1219 | |||
AK031556.1 | - | 5 | 1365 | |||
AK037407.1 | - | 5 | 1447 | |||
AK052190.1 | - | 5 | 1640 | |||
AK098107.1 | - | 5 | 1362 | |||
BC008534.1 | - | 4 | 796 | |||
BC008535.1 | - | 4 | 796 | |||
BC022633.1 | - | 2 | 66 | |||
BC071207.1 | - | 2 | 134 | |||
BC096483.1 | - | 5 | 1321 | 5310588 | ||
BC138234.1 | - | 5 | 1410 | 8861253 | ||
BC138235.1 | - | 5 | 1410 | 8861254 | ||
JX869941.1 | - | 5 | 1647 |