Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029537.1 | NP_083813.1 | 5, 6 | 520 | ||
GenBank mRNA | AK018332.1 | - | 4, 5 | 315 | ||
AK148132.1 | - | 5, 6 | 519 | |||
AK153960.1 | - | 5, 6 | 510 | |||
BC011208.1 | - | 5, 6 | 520 | 4163101 |