Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153596.3 | NP_705824.1 | 4 | 624 | ||
GenBank mRNA | BC030386.1 | - | 4 | 625 | 5400918 |