Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qsox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024945.1 | NP_001020116.1 | 12 | 1662 | ||
NM_023268.2 | NP_075757.1 | 12 | 1662 | |||
GenBank mRNA | AB044284.1 | - | 12 | 1570 | ||
AK004880.1 | - | 12 | 1608 | |||
AK012943.1 | - | 2 | 207 | |||
AK083938.1 | - | 12 | 1613 | |||
AK166839.1 | - | 12 | 1608 | |||
AK169877.1 | - | 12 | 1615 | |||
AK170449.1 | - | 12 | 1575 | |||
BC034131.1 | - | 9 | 1154 | |||
BC076590.1 | - | 12 | 1616 | 30548238 |