Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qsox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024945.1 | NP_001020116.1 | 4 | 538 | ||
NM_023268.2 | NP_075757.1 | 4 | 538 | |||
GenBank mRNA | AB044284.1 | - | 4 | 446 | ||
AK004880.1 | - | 4 | 484 | |||
AK083938.1 | - | 4 | 489 | |||
AK166839.1 | - | 4 | 484 | |||
AK169877.1 | - | 4 | 491 | |||
AK169920.1 | - | 4 | 487 | |||
AK170449.1 | - | 4 | 451 | |||
BC034131.1 | - | 1 | 30 | |||
BC076590.1 | - | 4 | 492 | 30548238 |