Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038695.3 | NP_001033784.2 | 13, 14 | 2135 | ||
NM_173001.3 | NP_766589.1 | 12, 13 | 2162 | |||
GenBank mRNA | AK129204.1 | - | 13, 14 | 2123 | ||
AK144825.1 | - | 12, 13 | 2163 | |||
AK166797.1 | - | 3, 4 | 558 | |||
BC031158.1 | - | 1, 2 | 222 | |||
BC031200.1 | - | 1, 2 | 206 | |||
BC032256.1 | - | 13, 14 | 2111 | |||
BC049876.1 | - | 6, 7 | 1205 | |||
BC059264.1 | - | 12, 13 | 2045 | 6409698 | ||
DQ323991.1 | - | 9, 10 | 1600 | |||
KJ461739.1 | - | 3, 4 | 466 |