Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166597.1 | NP_001160069.1 | 6, 7 | 1217 | ||
NM_138306.2 | NP_612179.2 | 5, 6 | 616 | |||
GenBank mRNA | AK087503.1 | - | 6, 7 | 1218 | ||
AK170936.1 | - | 2, 3 | 239 | |||
AK210058.1 | - | 1, 2 | 28 | |||
BC016217.1 | - | 5, 6 | 1563 | |||
BC049228.1 | - | 5, 6 | 652 | 3668704 |