Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npffr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133192.3 | NP_573455.2 | 4 | 1090 | ||
GenBank mRNA | AF236084.1 | - | 3 | 981 | ||
AF330054.1 | - | 3 | 975 | |||
AK050939.1 | - | 4 | 1091 |
Set Membership: |
G-protein coupled receptor |