Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rexo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024233.3 | NP_077195.2 | 1, 2 | 206 | ||
GenBank mRNA | AK007731.1 | - | 1, 2 | 208 | ||
AK090202.1 | - | 1, 2 | 210 | |||
AK150287.1 | - | 1, 2 | 122 | |||
AK171619.1 | - | 1, 2 | 109 | |||
AK172641.1 | - | 1, 2 | 184 | |||
AK178028.1 | - | 1, 2 | 48 | |||
AK209299.1 | - | 1, 2 | 48 | |||
AK211686.1 | - | 1, 2 | 95 | |||
AK212689.1 | - | 1, 2 | 95 | |||
BC003445.1 | - | 1, 2 | 142 | 2651110 |