Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rexo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024233.3 | NP_077195.2 | 5 | 562 | ||
GenBank mRNA | AK007731.1 | - | 5 | 564 | ||
AK150287.1 | - | 5 | 478 | |||
AK171619.1 | - | 5 | 465 | |||
AK172641.1 | - | 5 | 540 | |||
AK182577.1 | - | 2 | 104 | |||
BC003445.1 | - | 5 | 498 | 2651110 |