Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mycbpap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170671.2 | NP_733771.2 | 8, 9 | 1046 | ||
GenBank mRNA | AK029929.1 | - | 7, 8 | 1222 | ||
AK166780.1 | - | 1, 2 | 194 | |||
BC027327.1 | - | 4, 5 | 407 | 3991131 |