Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029842.1 | NP_001025013.1 | 5 | 1023 | ||
NM_134038.2 | NP_598799.1 | 5 | 966 | |||
GenBank mRNA | AK032026.1 | - | 5 | 1026 | ||
AK141927.1 | - | 1 | 1462 | |||
AK170472.1 | - | 5 | 889 | |||
BC017129.1 | - | 5 | 957 | 4506255 |