Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178911.4 | NP_849242.1 | 7 | 858 | ||
GenBank mRNA | AK045459.1 | - | 7 | 817 | ||
AK079549.1 | - | 7 | 817 | |||
AK079878.1 | - | 7 | 860 | |||
AK151551.1 | - | 7 | 1092 | |||
AK152969.1 | - | 6 | 705 | |||
AK170362.1 | - | 7 | 815 | |||
BC058565.1 | - | 7 | 808 | 6442038 |