Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipas39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142580.1 | NP_001136052.1 | 8, 9 | 723 | ||
NM_001142581.1 | NP_001136053.1 | 7, 8 | 666 | |||
NM_134044.3 | NP_598805.2 | 7, 8 | 680 | |||
GenBank mRNA | AK038165.1 | - | 8, 9 | 691 | ||
AK046322.1 | - | 8, 9 | 694 | |||
AK049643.1 | - | 8, 9 | 670 | |||
AK079055.1 | - | 8, 9 | 670 | |||
AK082261.1 | - | 8, 9 | 651 | |||
AK145043.1 | - | 706 | ||||
BC016646.1 | - | 7, 8 | 589 | 2651233 | ||
BC023716.1 | - | 7, 8 | 691 | 5346489 |