Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081043.1 | NP_001074512.1 | 20 | 2976 | ||
GenBank mRNA | AK173178.1 | - | 1 | 1022 | ||
BC059902.1 | - | 6 | 1705 |