Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289572.1 | NP_001276501.1 | 11 | 1571 | ||
NM_001289573.1 | NP_001276502.1 | 10 | 1166 | |||
NM_001289574.1 | NP_001276503.1 | 12 | 1733 | |||
NM_178914.4 | NP_849245.2 | 12 | 1667 | |||
GenBank mRNA | AK036359.1 | - | 12 | 1651 | ||
AK077500.1 | - | 12 | 1578 | |||
AK153698.1 | - | 10 | 1142 | |||
AK161308.1 | - | 11 | 1517 | |||
BC046960.1 | - | 12 | 1625 | 6391450 | ||
DQ196101.1 | - | 11 | 1397 |