Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028354.4 | NP_082630.2 | 9, 10 | 1179 | ||
GenBank mRNA | AK013427.1 | - | 8, 9 | 654 | ||
AK014855.1 | - | 9, 10 | 1159 | |||
AK033868.1 | - | 9, 10 | 1177 | |||
AK189362.1 | - | 3, 4 | 296 | |||
BC019804.1 | - | 6, 7 | 503 | 4018534 | ||
BC065162.1 | - | 8, 9 | 1149 | 6402921 |