Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042591.1 | NP_001036056.1 | 5 | 980 | ||
GenBank mRNA | AK084951.1 | - | 5 | 991 | ||
AK088288.1 | - | 5 | 995 | |||
AK122499.1 | - | 5 | 967 | |||
AK163364.1 | - | 5 | 525 | |||
BC023838.1 | - | 2 | 198 | |||
BC031125.1 | - | 3 | 358 | |||
BC044762.1 | - | 5 | 978 | |||
BC054844.1 | - | 5 | 980 | 6493441 | ||
BC098474.1 | - | 5 | 974 | 30094818 | ||
DQ861994.1 | - | 5 | 725 |