Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epdr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134065.4 | NP_598826.3 | 3 | 771 | ||
GenBank mRNA | AF353717.1 | - | 3 | 584 | ||
AK034083.1 | - | 3 | 740 | |||
AK038733.1 | - | 3 | 746 | |||
AK046185.1 | - | 3 | 746 | |||
AK160413.1 | - | 3 | 706 | |||
AK166252.1 | - | 3 | 712 | |||
BC014708.1 | - | 3 | 726 | 4223134 | ||
DQ914440.1 | - | 2 | 294 |