Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phyhip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145981.3 | NP_666093.1 | 2, 3 | 463 | ||
GenBank mRNA | AK049900.1 | - | 2, 3 | 464 | ||
BC030494.1 | - | 2, 3 | 460 | |||
BC139360.1 | - | 3, 4 | 446 | 8862382 |