Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253813.1 | NP_001240742.1 | 18 | 2324 | ||
NM_175344.4 | NP_780553.2 | 17 | 2261 | |||
GenBank mRNA | AK089300.1 | - | 18 | 2397 | ||
AK159411.1 | - | 18 | 2273 | |||
BC060732.1 | - | 17 | 2261 | 6409332 |