Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166456.1 | NP_001159928.1 | 4 | 530 | ||
NM_001166458.1 | NP_001159930.1 | 4 | 737 | |||
NM_134086.4 | NP_598847.2 | 4 | 1223 | |||
GenBank mRNA | AF184240.2 | - | 3 | 315 | ||
AK029189.1 | - | 4 | 400 | |||
AK034130.1 | - | 4 | 1225 | |||
AK035098.1 | - | 3 | 242 | |||
AK047459.1 | - | 4 | 754 | |||
AK049173.1 | - | 4 | 437 | |||
AK049294.1 | - | 4 | 503 | |||
AK050914.1 | - | 4 | 438 | |||
AK081724.1 | - | 4 | 739 | |||
AK082683.1 | - | 4 | 438 | |||
AK162612.1 | - | 4 | 588 | |||
AK165188.1 | - | 4 | 790 | |||
BC030378.1 | - | 4 | 439 | 5375319 |