Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168250.1 | NP_001161722.1 | 5, 6 | 703 | ||
NM_134092.3 | NP_598853.3 | 5, 6 | 703 | |||
GenBank mRNA | AJ278508.1 | - | 5, 6 | 624 | ||
AK080036.1 | - | 5, 6 | 517 | |||
AK088549.1 | - | 5, 6 | 517 | |||
AK133855.1 | - | 5, 6 | 551 | |||
BC052197.1 | - | 5, 6 | 703 | 6511752 |