Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178919.4 | NP_849250.1 | 2, 3 | 565 | ||
GenBank mRNA | AK042427.1 | - | 2, 3 | 524 | ||
AK084606.1 | - | 2, 3 | 391 | |||
AK085435.1 | - | 2, 3 | 516 | |||
AK149916.1 | - | 2, 3 | 517 | |||
AK149934.1 | - | 2, 3 | 566 | |||
AK171488.1 | - | 2, 3 | 546 | |||
BC038039.1 | - | 1, 2 | 1067 | |||
BC079532.1 | - | 2, 3 | 461 | 6403184 |