Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Espl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014976.2 | NP_001014976.1 | 6 | 1521 | ||
GenBank mRNA | AK129072.1 | - | 6 | 1488 | ||
AY588413.1 | - | 6 | 1459 | |||
BC082603.1 | - | 6 | 1521 | 30668743 | ||
BC145844.1 | - | 6 | 1492 | 40131147 | ||
BC145846.1 | - | 6 | 1438 | 40131149 |