Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Topors | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134097.3 | NP_598858.2 | 3 | 2572 | ||
GenBank mRNA | AB072395.1 | - | 3 | 2529 | ||
AB104865.1 | - | 3 | 2557 | |||
AK134075.1 | - | 3 | 2568 | |||
BC019421.1 | - | 3 | 2530 | |||
BC037141.1 | - | 3 | 2550 | |||
BC040797.1 | - | 3 | 2560 | 3492178 |