Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145931.2 | NP_666043.2 | 12 | 1445 | ||
GenBank mRNA | AK028438.1 | - | 12 | 1373 | ||
AK040507.1 | - | 12 | 1393 | |||
AK049834.1 | - | 12 | 1380 | |||
AK052830.1 | - | 13 | 1556 | |||
AK150952.1 | - | 12 | 1364 | |||
AK166876.1 | - | 13 | 1533 | |||
BC027330.1 | - | 12 | 1341 | 3674968 | ||
BC029320.1 | - | 12 | 1343 |